Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291494.1 | NP_001278423.1 | 3 | 948 | ||
NM_003638.2 | NP_003629.2 | 3 | 948 | |||
XM_011519752.2 | XP_011518054.1 | 3 | 942 | |||
GenBank mRNA | AK297740.1 | - | 3 | 460 | ||
AK308240.1 | - | 3 | 465 | |||
L36531.1 | - | 3 | 270 |