Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000314.6 | NP_000305.3 | 9 | 2695 | ||
NM_001304717.2 | NP_001291646.2 | 9 | 2695 | |||
NM_001304718.1 | NP_001291647.1 | 9 | 2809 | |||
GenBank mRNA | U92436.1 | - | 9 | 2698 |