Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000070.2 | NP_000061.1 | 17 | 2278 | ||
NM_024344.1 | NP_077320.1 | 16 | 2260 | |||
NM_173087.1 | NP_775110.1 | 14 | 2002 | |||
NM_173088.1 | NP_775111.1 | 6 | 605 | |||
NM_173089.1 | NP_775112.1 | 2 | 235 | |||
NM_173090.1 | NP_775113.1 | 3 | 253 | |||
GenBank mRNA | AF127764.1 | - | 16 | 2175 | ||
AF127765.3 | - | 14 | 1917 | |||
AK027249.1 | - | 1 | 895 | |||
AK308103.1 | - | 17 | 2278 | |||
BC003169.1 | - | 6 | 605 | |||
BC003521.1 | - | 2 | 217 | |||
BC004883.2 | - | 2 | 235 | |||
BC007810.2 | - | 3 | 201 | |||
BC067126.1 | - | 2 | 218 | |||
BC100782.1 | - | 2 | 216 | |||
BC100783.1 | - | 2 | 216 | |||
BC100784.2 | - | 2 | 216 | |||
BC107791.1 | - | 3 | 368 | |||
BC128605.1 | - | 2 | 216 | |||
BC146649.1 | - | 17 | 2191 | 40147158 | ||
BC146672.1 | - | 17 | 2191 | 40148444 | ||
BT007322.1 | - | 5 | 436 | |||
EU791850.1 | - | 17 | 1711 | |||
EU791851.1 | - | 16 | 1567 | |||
KU177954.1 | - | 5 | 436 | |||
KU177955.1 | - | 5 | 537 | |||
KU177956.1 | - | 3 | 304 | |||
X85030.1 | - | 17 | 1972 |