Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000070.2 | NP_000061.1 | 22 | 2641 | ||
NM_024344.1 | NP_077320.1 | 21 | 2623 | |||
NM_173087.1 | NP_775110.1 | 19 | 2365 | |||
NM_173088.1 | NP_775111.1 | 11 | 968 | |||
NM_173089.1 | NP_775112.1 | 7 | 598 | |||
NM_173090.1 | NP_775113.1 | 8 | 616 | |||
GenBank mRNA | AF127764.1 | - | 21 | 2538 | ||
AF127765.3 | - | 19 | 2280 | |||
AK027249.1 | - | 6 | 1258 | |||
BC003169.1 | - | 11 | 968 | |||
BC003521.1 | - | 7 | 580 | |||
BC004883.2 | - | 7 | 598 | |||
BC007810.2 | - | 8 | 564 | |||
BC067126.1 | - | 7 | 581 | |||
BC100782.1 | - | 7 | 579 | |||
BC100783.1 | - | 7 | 579 | |||
BC100784.2 | - | 7 | 579 | |||
BC107791.1 | - | 8 | 731 | |||
BC128605.1 | - | 7 | 579 | |||
BC146649.1 | - | 22 | 2554 | 40147158 | ||
BC146672.1 | - | 22 | 2554 | 40148444 | ||
BT007322.1 | - | 10 | 799 | |||
EU791850.1 | - | 22 | 2074 | |||
EU791851.1 | - | 21 | 1930 | |||
KU177954.1 | - | 10 | 799 | |||
KU177955.1 | - | 10 | 900 | |||
KU177956.1 | - | 8 | 667 | |||
X85030.1 | - | 22 | 2335 |