Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130145.2 | NP_001123617.1 | 9 | 1699 | ||
NM_001195044.1 | NP_001181973.1 | 8 | 1651 | |||
NM_001195045.1 | NP_001181974.1 | 9 | 1296 | |||
NM_001282097.1 | NP_001269026.1 | 8 | 1585 | |||
NM_001282098.1 | NP_001269027.1 | 7 | 1549 | |||
NM_001282099.1 | NP_001269028.1 | 8 | 1597 | |||
NM_001282100.1 | NP_001269029.1 | 8 | 1663 | |||
NM_001282101.1 | NP_001269030.1 | 9 | 1711 | |||
NM_006106.4 | NP_006097.2 | 7 | 1537 | |||
XM_005271378.3 | XP_005271435.1 | 9 | 2221 | |||
XM_005271380.3 | XP_005271437.1 | 9 | 2209 | |||
XM_005271381.3 | XP_005271438.1 | 8 | 2173 | |||
XM_005271383.3 | XP_005271440.1 | 8 | 2161 | |||
XM_011542555.2 | XP_011540857.1 | 9 | 1309 | |||
XM_011542556.2 | XP_011540858.1 | 9 | 1303 | |||
XM_017017093.1 | XP_016872582.1 | 8 | 1243 | |||
GenBank mRNA | AB567720.1 | - | 8 | 1197 | ||
AB567721.1 | - | 9 | 1311 | |||
AK300414.1 | - | 9 | 1014 | |||
AK304485.1 | - | 6 | 572 | |||
AK310620.1 | - | 3 | 259 | |||
AK316116.1 | - | 9 | 1222 | |||
AL832620.2 | - | 8 | 992 | |||
AY316529.1 | - | 8 | 1263 | |||
BC026212.2 | - | 3 | 280 | |||
BC038235.1 | - | 9 | 1604 | 5747370 |