Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000083.2 | NP_000074.2 | 15, 16 | 1879 | ||
NR_046453.1 | 14, 15 | 1732 | ||||
XM_011515782.2 | XP_011514084.1 | 5, 6 | 627 | |||
XM_017011739.1 | XP_016867228.1 | 12, 13 | 1604 | |||
XM_017011740.1 | XP_016867229.1 | 12, 13 | 1580 | |||
GenBank mRNA | BC112156.1 | - | 15, 16 | 1844 | 8327624 | |
BC113495.1 | - | 15, 16 | 1844 | 8322547 | ||
BC143796.1 | - | 14, 15 | 1697 | |||
M97820.1 | - | 12, 13 | 1283 | |||
Z25884.1 | - | 15, 16 | 1879 |