Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000083.2 | NP_000074.2 | 16, 17 | 2010 | ||
NR_046453.1 | 15, 16 | 1863 | ||||
XM_011515782.2 | XP_011514084.1 | 6, 7 | 758 | |||
XM_017011739.1 | XP_016867228.1 | 13, 14 | 1735 | |||
XM_017011740.1 | XP_016867229.1 | 13, 14 | 1711 | |||
GenBank mRNA | BC112156.1 | - | 16, 17 | 1975 | 8327624 | |
BC113495.1 | - | 16, 17 | 1975 | 8322547 | ||
BC143796.1 | - | 15, 16 | 1828 | |||
M97820.1 | - | 13, 14 | 1414 | |||
Z25884.1 | - | 16, 17 | 2010 |