Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 33, 34 | 4945 | ||
NM_001289397.1 | NP_001276326.1 | 32, 33 | 4730 | |||
NM_001289398.1 | NP_001276327.1 | 6, 7 | 912 | |||
NM_001289399.1 | NP_001276328.1 | 7, 8 | 899 | |||
NM_001289400.1 | NP_001276329.1 | 7, 8 | 1320 | |||
NM_003070.4 | NP_003061.3 | 33, 34 | 4958 | |||
NM_139045.3 | NP_620614.2 | 32, 33 | 4904 | |||
GenBank mRNA | AK094076.1 | - | 7, 8 | 1320 | ||
AK298045.1 | - | 7, 8 | 899 | |||
AK300093.1 | - | 8, 9 | 964 | |||
AK308941.1 | - | 6, 7 | 1057 | |||
AL558829.3 | - | 8, 9 | 904 | |||
CD107405.1 | - | 5, 6 | 667 | |||
CD513892.1 | - | 5, 6 | 708 | |||
X72889.1 | - | 33, 34 | 4946 |