Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPY19L2 SIRNA Assays ›
Gene Symbol:
DPY19L2
Gene Name:
dpy-19 like 2
Gene Aliases:
SPATA34, SPGF9
Chromosome Location:
Chr.12: 63558913 - 63669201 on Build GRCh38
UniGene ID:
Hs.533644
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s536711
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPY19L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_173812.4
NP_776173.3
22
2526
XM_006719352.2
XP_006719415.1
22
2452
XM_011538215.2
XP_011536517.1
20
1992
XM_011538218.2
XP_011536520.1
15
1747
XM_017019188.1
XP_016874677.1
22
2573
XM_017019189.1
XP_016874678.1
21
2513
XM_017019190.1
XP_016874679.1
21
2447
XM_017019191.1
XP_016874680.1
20
2423
XM_017019192.1
XP_016874681.1
20
2411
XM_017019193.1
XP_016874682.1
21
2108
XM_017019194.1
XP_016874683.1
21
2135
XM_017019195.1
XP_016874684.1
22
2183
XM_017019196.1
XP_016874685.1
20
2272
XM_017019197.1
XP_016874686.1
21
2406
XM_017019198.1
XP_016874687.1
18
2127
XM_017019199.1
XP_016874688.1
18
2196
XM_017019201.1
XP_016874690.1
21
2369
XM_017019202.1
XP_016874691.1
20
1982
XM_017019203.1
XP_016874692.1
20
2204
XM_017019204.1
XP_016874693.1
21
2442
XM_017019205.1
XP_016874694.1
18
2296
GenBank mRNA
AK057511.1
-
22
2497
AK303727.1
-
6
987
AL833344.1
-
22
2453
AY358792.1
-
22
2447
BC031225.1
-
16
1528
BC068442.1
-
22
2529
5297524
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
mannosyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0