Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000510.2 | NP_000501.1 | 3 | 291 | ||
NM_001018080.1 | NP_001018090.1 | 3 | 261 | |||
GenBank mRNA | BC113488.1 | - | 2 | 252 | 8322540 | |
BC113490.1 | - | 2 | 252 | 8322542 | ||
EF585489.1 | - | 2 | 168 |