Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318521.1 | NP_001305450.1 | 14 | 1490 | ||
NM_001318523.1 | NP_001305452.1 | 12 | 1327 | |||
NM_006733.3 | NP_006724.2 | 13 | 1584 | |||
XM_005262111.2 | XP_005262168.1 | 14 | 1454 | |||
XM_011530894.2 | XP_011529196.1 | 14 | 1483 | |||
XM_011530895.2 | XP_011529197.1 | 15 | 1770 | |||
XM_011530897.2 | XP_011529199.1 | 15 | 1645 | |||
XM_011530899.2 | XP_011529201.1 | 13 | 1451 | |||
XM_017029383.1 | XP_016884872.1 | 14 | 1483 | |||
GenBank mRNA | AK302986.1 | - | 14 | 1483 | ||
BC012462.2 | - | 12 | 1328 | 4537558 | ||
X97249.1 | - | 13 | 1583 |