Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012415.2 | NP_001012415.2 | 4 | 452 | ||
NM_001101677.1 | NP_001095147.1 | 4 | 452 | |||
XM_005263403.3 | XP_005263460.1 | 4 | 403 | |||
XM_006717109.3 | XP_006717172.1 | 5 | 1528 | |||
XM_011518698.2 | XP_011517000.1 | 6 | 1557 | |||
GenBank mRNA | AY884306.1 | - | 4 | 397 | ||
AY902244.1 | - | 4 | 452 | |||
BC031861.1 | - | 4 | 463 | 4826967 | ||
EB710265.1 | - | 2 | 149 |