Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017747.2 | NP_060217.1 | 29 | 5760 | ||
GenBank mRNA | AF521882.1 | - | 29 | 5666 | ||
AK024189.1 | - | 6 | 1384 | |||
AL136943.1 | - | 4 | 896 | |||
BC040231.1 | - | 4 | 957 |
Gene Symbol: ANKHD1-EIF4EBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020690.5 | NP_065741.3 | 29 | 5760 | ||
GenBank mRNA | AF521883.1 | - | 29 | 5666 | ||
BC144623.1 | - | 29 | 5794 |