Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207581.3 | NP_997464.2 | 6 | 1217 | ||
XM_017022180.1 | XP_016877669.1 | 6 | 1272 | |||
GenBank mRNA | BC137465.1 | - | 6 | 1138 | 9021465 | |
BX537581.1 | - | 6 | 1501 | |||
DQ489734.1 | - | 6 | 1068 |
Gene Symbol: DUOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB850544.1 | - | 1 | 534 |