Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252216.1 | NP_001239145.1 | 3 | 372 | ||
NM_213609.3 | NP_998774.2 | 3 | 646 | |||
XM_017006386.1 | XP_016861875.1 | 2 | 793 | |||
XM_017006387.1 | XP_016861876.1 | 3 | 1285 | |||
XM_017006388.1 | XP_016861877.1 | 3 | 1078 | |||
XM_017006389.1 | XP_016861878.1 | 2 | 790 | |||
XM_017006390.1 | XP_016861879.1 | 3 | 1286 | |||
GenBank mRNA | AK289668.1 | - | 3 | 635 | ||
AK290446.1 | - | 3 | 568 | |||
AY325114.1 | - | 3 | 372 | |||
BC025746.1 | - | 3 | 548 | 5200142 | ||
DA485098.1 | - | 3 | 574 |