Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304464.1 | NP_001291393.1 | 4 | 285 | ||
NM_003094.3 | NP_003085.1 | 4 | 241 | |||
NR_130746.1 | 3 | 244 | ||||
GenBank mRNA | AA344740.1 | - | 3 | 191 | ||
AI263891.1 | - | 3 | 220 | |||
AK307255.1 | - | 4 | 271 | |||
AK312130.1 | - | 4 | 218 | |||
BC002639.2 | - | 4 | 210 | 3609230 | ||
BC072433.1 | - | 4 | 189 | |||
BC090951.1 | - | 4 | 214 | 5204266 | ||
CN272809.1 | - | 3 | 169 | |||
M15919.1 | - | 4 | 134 | |||
M37716.1 | - | 4 | 211 | |||
X12466.1 | - | 4 | 211 |