Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077665.2 | NP_001071133.2 | 7 | 951 | ||
XM_011539815.2 | XP_011538117.1 | 4 | 176 | |||
XM_017016268.1 | XP_016871757.1 | 7 | 672 | |||
XM_017016269.1 | XP_016871758.1 | 7 | 672 | |||
XM_017016270.1 | XP_016871759.1 | 6 | 906 | |||
XM_017016271.1 | XP_016871760.1 | 9 | 674 | |||
XM_017016272.1 | XP_016871761.1 | 8 | 604 | |||
XM_017016273.1 | XP_016871762.1 | 8 | 1297 | |||
XM_017016274.1 | XP_016871763.1 | 4 | 127 |
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC171820.1 | - | 7 | 664 |