Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004959.4 | NP_004950.2 | 7 | 1337 | ||
XM_005251871.3 | XP_005251928.1 | 7 | 1365 | |||
XM_011518455.2 | XP_011516757.1 | 7 | 1215 | |||
GenBank mRNA | AB307718.1 | - | 6 | 1150 | ||
AK090858.1 | - | 2 | 783 | |||
BC032501.1 | - | 7 | 1337 | 5227690 | ||
HQ692856.1 | - | 6 | 1150 | |||
HQ709184.1 | - | 5 | 1030 | |||
U76388.1 | - | 6 | 1165 |