Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001551.3 | NP_001001551.2 | 5 | 539 | ||
NM_001256915.1 | NP_001243844.1 | 5 | 713 | |||
NR_046421.1 | 4 | 508 | ||||
NR_046422.1 | 6 | 1125 | ||||
XM_006717110.3 | XP_006717173.1 | 5 | 658 | |||
XM_006717111.3 | XP_006717174.1 | 5 | 3067 | |||
XM_017014719.1 | XP_016870208.1 | 3 | 464 | |||
XM_017014724.1 | XP_016870213.1 | 5 | 1370 | |||
XM_017014725.1 | XP_016870214.1 | 5 | 499 | |||
GenBank mRNA | AF086281.1 | - | 1 | 116 | ||
AY615889.1 | - | 5 | 430 | |||
BC036421.1 | - | 6 | 1128 | |||
BC045782.1 | - | 6 | 1123 | |||
BC142991.1 | - | 6 | 630 | 8860441 | ||
CA866332.1 | - | 4 | 417 |