Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001712.2 | NP_001001712.2 | 5 | 543 | ||
GenBank mRNA | AK091788.1 | - | 2 | 1031 | ||
AL832485.1 | - | 1 | 784 | |||
AY301271.1 | - | 4 | 448 | |||
BC036875.1 | - | 2 | 145 | |||
BC040963.1 | - | 5 | 1788 | |||
BC046377.1 | - | 1 | 73 | |||
BC133045.1 | - | 5 | 543 | 40147067 | ||
BC144440.1 | - | 3 | 325 | |||
BC144441.1 | - | 4 | 435 | 9052964 | ||
BC171741.1 | - | 3 | 263 |