Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001794.3 | NP_001001794.3 | 20 | 1767 | ||
XM_005261914.3 | XP_005261971.1 | 21 | 1942 | |||
XM_011530692.2 | XP_011528994.1 | 21 | 1957 | |||
XM_011530693.2 | XP_011528995.1 | 20 | 1736 | |||
XM_017028801.1 | XP_016884290.1 | 19 | 1778 | |||
GenBank mRNA | AK054743.1 | - | 17 | 2214 | ||
BC031069.1 | - | 20 | 1767 | 5296726 |