Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001794.3 | NP_001001794.3 | 12 | 1087 | ||
XM_005261914.3 | XP_005261971.1 | 13 | 1262 | |||
XM_011530692.2 | XP_011528994.1 | 13 | 1262 | |||
XM_011530693.2 | XP_011528995.1 | 12 | 1041 | |||
XM_017028801.1 | XP_016884290.1 | 11 | 1098 | |||
GenBank mRNA | AK054743.1 | - | 10 | 1206 | ||
BC031069.1 | - | 12 | 1087 | 5296726 |