Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002036.3 | NP_001002036.3 | 2 | 166 | ||
XM_011511205.2 | XP_011509507.1 | 1 | 981 | |||
XM_011511206.2 | XP_011509508.1 | 2 | 1204 | |||
XM_011511207.2 | XP_011509509.1 | 1 | 982 | |||
XM_011511208.2 | XP_011509510.1 | 1 | 982 | |||
GenBank mRNA | AJ537600.2 | - | 2 | 166 | ||
BC107126.1 | - | 2 | 165 | |||
BC107127.1 | - | 2 | 166 | 40023980 |