Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000147.4 | NP_000138.2 | 6 | 1102 | ||
XM_005245821.2 | XP_005245878.1 | 6 | 1239 | |||
XM_011541167.2 | XP_011539469.1 | 6 | 687 | |||
XM_017000905.1 | XP_016856394.1 | 6 | 784 | |||
GenBank mRNA | AB074175.1 | - | 6 | 1023 | ||
AK092914.1 | - | 7 | 957 | |||
AK095443.1 | - | 7 | 974 | |||
AK095455.1 | - | 7 | 988 | |||
AK225468.1 | - | 6 | 1023 | |||
AK314649.1 | - | 6 | 1023 | |||
BC017338.2 | - | 6 | 1025 | 4871788 | ||
M10355.1 | - | 6 | 797 | |||
M29877.1 | - | 6 | 1022 | |||
X01390.1 | - | 6 | 797 |