Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1L6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040044.1 | 4 | 535 | |||
GenBank mRNA | AK093040.1 | - | 4 | 535 | ||
BC137357.1 | - | 4 | 502 | 9021355 |