Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC440040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027044.1 | 4 | 1322 | |||
GenBank mRNA | AK294678.1 | - | 4 | 1322 | ||
BC141819.1 | - | 4 | 1230 | |||
BC142657.1 | - | 4 | 1230 |