Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033399.1 | 3 | 403 |
Gene Symbol: LOC642846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024374.1 | 3 | 452 | |||
GenBank mRNA | AK296420.1 | - | 3 | 460 | ||
AK303618.1 | - | 3 | 230 | |||
AK309820.1 | - | 3 | 320 |