Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000148.3 | NP_000139.1 | 4 | 1506 | ||
XM_006723127.2 | XP_006723190.1 | 3 | 1498 | |||
XM_017026552.1 | XP_016882041.1 | 3 | 1418 | |||
XM_017026553.1 | XP_016882042.1 | 4 | 1332 | |||
GenBank mRNA | BC074732.2 | - | 2 | 572 | 30915185 | |
M35531.1 | - | 2 | 634 |