Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080534.1 | NP_001074003.1 | 1 | 2785 | ||
XM_005254394.4 | XP_005254451.1 | 2 | 3736 | |||
XM_017022220.1 | XP_016877709.1 | 6 | 3656 | |||
XM_017022221.1 | XP_016877710.1 | 5 | 3536 | |||
XM_017022222.1 | XP_016877711.1 | 4 | 3434 | |||
XM_017022223.1 | XP_016877712.1 | 3 | 8968 | |||
XM_017022224.1 | XP_016877713.1 | 2 | 3668 | |||
XM_017022225.1 | XP_016877714.1 | 1 | 1175 |