Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000511.5 | NP_000502.4 | 2 | 246 | ||
NM_001097638.2 | NP_001091107.1 | 2 | 236 | |||
GenBank mRNA | AK290277.1 | - | 2 | 232 | ||
AY751739.1 | - | 1 | 125 | |||
BC001899.1 | - | 2 | 186 | |||
BC121066.1 | - | 2 | 168 | |||
BE266792.1 | - | 2 | 168 | |||
BT006657.1 | - | 1 | 125 | |||
D87942.1 | - | 2 | 236 | |||
DA918525.1 | - | 2 | 236 |