Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004341.2 | NP_001004341.1 | 2, 3 | 588 | ||
GenBank mRNA | AK131392.1 | - | 2, 3 | 588 |