Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004342.3 | NP_001004342.3 | 10 | 2352 | ||
NM_001300889.1 | NP_001287818.1 | 12 | 2166 | |||
XM_011544192.2 | XP_011542494.1 | 10 | 2346 | |||
GenBank mRNA | AY253917.1 | - | 12 | 2124 |