Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000150.2 | NP_000141.1 | 3 | 2814 | ||
NM_001040701.1 | NP_001035791.1 | 2 | 2686 | |||
XM_005259526.4 | XP_005259583.1 | 4 | 2881 | |||
XM_011527868.2 | XP_011526170.1 | 4 | 2953 | |||
XM_011527869.2 | XP_011526171.1 | 4 | 2950 | |||
XM_011527870.2 | XP_011526172.1 | 5 | 3061 | |||
XM_011527872.2 | XP_011526174.1 | 5 | 3081 | |||
XM_011527873.2 | XP_011526175.1 | 4 | 3016 | |||
XM_011527874.2 | XP_011526176.1 | 4 | 3036 | |||
XM_011527875.2 | XP_011526177.1 | 5 | 3107 | |||
XM_011527876.2 | XP_011526178.1 | 4 | 3062 | |||
XM_011527878.2 | XP_011526180.1 | 4 | 2953 | |||
XM_011527879.2 | XP_011526181.1 | 3 | 2908 | |||
GenBank mRNA | AF131211.1 | - | 4 | 1895 | ||
AK098073.1 | - | 3 | 1671 | |||
BC061700.1 | - | 2 | 2676 | 4606746 | ||
U27333.1 | - | 3 | 2769 | |||
U27334.1 | - | 3 | 2092 | |||
U27335.1 | - | 2 | 2644 | |||
U27336.1 | - | 2 | 1967 | |||
U27337.1 | - | 3 | 2254 |