Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM150C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080506.1 | NP_001073975.1 | 6 | 428 | ||
GenBank mRNA | AK297434.1 | - | 6 | 555 | ||
AK297512.1 | - | 6 | 433 | |||
AK299942.1 | - | 6 | 418 | |||
AK301116.1 | - | 6 | 402 | |||
BC147019.1 | - | 6 | 411 | 9056947 | ||
BC147027.1 | - | 6 | 411 | 9056955 |
Gene Symbol: RPS6P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052613.1 | - | 1 | 198 |