Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282601.1 | NP_001269530.1 | 4 | 620 | ||
NM_001282602.1 | NP_001269531.1 | 4 | 620 | |||
NM_001282604.1 | NP_001269533.1 | 5 | 669 | |||
NM_001282605.1 | NP_001269534.1 | 4 | 620 | |||
NM_006601.6 | NP_006592.3 | 4 | 620 | |||
NR_104219.1 | 4 | 957 | ||||
XM_005268576.4 | XP_005268633.1 | 4 | 2210 | |||
XM_006719199.2 | XP_006719262.1 | 4 | 969 | |||
XM_011537773.2 | XP_011536075.1 | 4 | 528 | |||
XM_011537774.2 | XP_011536076.1 | 4 | 969 | |||
XM_017018716.1 | XP_016874205.1 | 4 | 969 | |||
GenBank mRNA | AK098214.1 | - | 4 | 957 | ||
AK291945.1 | - | 4 | 374 | |||
AK298147.1 | - | 4 | 496 | |||
AK298160.1 | - | 4 | 434 | |||
AL603017.1 | - | 4 | 506 | |||
BC003005.1 | - | 4 | 317 | 2821965 | ||
BM541503.1 | - | 5 | 486 | |||
BU167210.1 | - | 4 | 475 | |||
L24804.1 | - | 4 | 443 |