Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282602.1 | NP_001269531.1 | 5 | 729 | ||
NM_001282603.1 | NP_001269532.1 | 5 | 720 | |||
NM_001282604.1 | NP_001269533.1 | 7 | 868 | |||
NM_006601.6 | NP_006592.3 | 6 | 819 | |||
NR_104219.1 | 6 | 1156 | ||||
XM_005268576.4 | XP_005268633.1 | 6 | 2409 | |||
XM_006719199.2 | XP_006719262.1 | 5 | 1078 | |||
XM_011537773.2 | XP_011536075.1 | 6 | 727 | |||
XM_017018716.1 | XP_016874205.1 | 6 | 1168 | |||
GenBank mRNA | AK098214.1 | - | 6 | 1156 | ||
AK291945.1 | - | 6 | 573 | |||
AK293139.1 | - | 5 | 678 | |||
AK295208.1 | - | 5 | 474 | |||
AK298160.1 | - | 5 | 543 | |||
BC003005.1 | - | 6 | 516 | 2821965 | ||
BC021167.1 | - | 2 | 70 | |||
BM541503.1 | - | 7 | 685 | |||
L24804.1 | - | 6 | 642 |