Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113541.1 | NP_001107013.1 | 4 | 3307 | ||
XM_011545671.2 | XP_011543973.1 | 4 | 3378 |
Gene Symbol: SPATA31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085452.2 | NP_001078921.2 | 4 | 3340 | ||
XM_011518006.1 | XP_011516308.1 | 4 | 3378 | |||
GenBank mRNA | AK302569.1 | - | 4 | 3340 | ||
BC146855.1 | - | 4 | 3299 | 9056783 |
Gene Symbol: SPATA31A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145196.1 | NP_001138668.1 | 4 | 3295 | ||
XM_011517871.2 | XP_011516173.1 | 4 | 3302 |
Gene Symbol: SPATA31A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015667.2 | NP_056482.2 | 4 | 3307 | ||
GenBank mRNA | BC040528.1 | - | 4 | 3307 | 4838470 |
Gene Symbol: SPATA31C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145124.1 | NP_001138596.1 | 4 | 2863 | ||
XM_011518701.2 | XP_011517003.1 | 5 | 3501 | |||
XM_011518702.1 | XP_011517004.1 | 4 | 3420 | |||
XM_011518703.1 | XP_011517005.1 | 4 | 3300 |
Gene Symbol: SPATA31C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166137.1 | NP_001159609.1 | 4 | 2845 | ||
GenBank mRNA | AK302448.1 | - | 4 | 2845 | ||
AL080179.1 | - | 1 | 347 |