Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002037.5 | NP_002028.1 | 8 | 1190 | ||
NM_153047.3 | NP_694592.1 | 5 | 594 | |||
NM_153048.3 | NP_694593.1 | 6 | 686 | |||
XM_005266890.3 | XP_005266947.1 | 8 | 771 | |||
XM_005266892.3 | XP_005266949.1 | 8 | 771 | |||
XM_017010650.1 | XP_016866139.1 | 7 | 928 | |||
XM_017010651.1 | XP_016866140.1 | 7 | 832 | |||
XM_017010652.1 | XP_016866141.1 | 6 | 842 | |||
XM_017010653.1 | XP_016866142.1 | 6 | 686 | |||
XM_017010654.1 | XP_016866143.1 | 6 | 667 | |||
XM_017010655.1 | XP_016866144.1 | 8 | 771 | |||
GenBank mRNA | AB451293.1 | - | 5 | 583 | ||
AB451426.1 | - | 5 | 583 | |||
AK056699.1 | - | 6 | 776 | |||
BC032496.1 | - | 6 | 637 | 5219761 | ||
CX872011.1 | - | 6 | 624 | |||
M14333.1 | - | 8 | 1162 | |||
M14676.1 | - | 8 | 953 | |||
S74774.1 | - | 5 | 583 |