Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002037.5 | NP_002028.1 | 12 | 1853 | ||
NM_153047.3 | NP_694592.1 | 9 | 1248 | |||
NM_153048.3 | NP_694593.1 | 9 | 1184 | |||
XM_005266890.3 | XP_005266947.1 | 12 | 1425 | |||
XM_005266892.3 | XP_005266949.1 | 11 | 1269 | |||
XM_017010650.1 | XP_016866139.1 | 11 | 1591 | |||
XM_017010651.1 | XP_016866140.1 | 11 | 1495 | |||
XM_017010652.1 | XP_016866141.1 | 10 | 1505 | |||
XM_017010653.1 | XP_016866142.1 | 10 | 1349 | |||
XM_017010654.1 | XP_016866143.1 | 10 | 1330 | |||
GenBank mRNA | AB451293.1 | - | 9 | 1246 | ||
AB451426.1 | - | 9 | 1246 | |||
AK056699.1 | - | 10 | 1430 | |||
BC032496.1 | - | 9 | 1135 | 5219761 | ||
M14333.1 | - | 12 | 1825 | |||
M14676.1 | - | 12 | 1616 | |||
S74774.1 | - | 9 | 1237 |