Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001466.3 | NP_001457.1 | 1 | 1809 | ||
GenBank mRNA | AB017364.1 | - | 1 | 1602 | ||
BC052266.1 | - | 1 | 1794 | |||
BC113402.1 | - | 2 | 1611 | 8322454 | ||
BC113404.1 | - | 2 | 1611 | 8322456 | ||
L37882.1 | - | 1 | 1696 |
Set Membership: |
G-protein coupled receptor |