Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033540.2 | NP_284941.2 | 16 | 1954 | ||
XM_005247596.3 | XP_005247653.2 | 16 | 1912 | |||
XM_011512963.2 | XP_011511265.1 | 13 | 1540 | |||
GenBank mRNA | AB043588.1 | - | 15 | 1919 | ||
AB043589.1 | - | 16 | 1919 | |||
AF054986.1 | - | 6 | 711 | |||
AF329637.1 | - | 15 | 1828 | |||
AK000700.1 | - | 15 | 1784 | |||
AK314306.1 | - | 16 | 1911 | |||
BC040557.1 | - | 16 | 1954 | 5270347 | ||
CR627379.1 | - | 13 | 1603 | |||
U95822.1 | - | 10 | 1084 |