Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002038.3 | NP_002029.3 | 5 | 571 | ||
NM_022872.2 | NP_075010.1 | 5 | 583 | |||
NM_022873.2 | NP_075011.1 | 5 | 595 | |||
GenBank mRNA | AK024814.1 | - | 5 | 551 | ||
BC011601.2 | - | 5 | 522 | 3140035 | ||
BC015603.2 | - | 5 | 524 | 4654295 | ||
X02492.1 | - | 5 | 517 |