Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145210.2 | NP_001138682.1 | 4 | 1787 | ||
NM_001243535.1 | NP_001230464.1 | 3 | 1246 | |||
NM_001243536.1 | NP_001230465.1 | 3 | 1227 | |||
XM_005244752.4 | XP_005244809.1 | 4 | 2683 | |||
XM_006710647.3 | XP_006710710.1 | 4 | 2702 | |||
XM_011541489.2 | XP_011539791.1 | 3 | 2777 | |||
XM_017001324.1 | XP_016856813.1 | 4 | 2009 | |||
XM_017001325.1 | XP_016856814.1 | 2 | 2236 | |||
GenBank mRNA | BC018095.1 | - | 3 | 1066 | ||
BC094869.1 | - | 3 | 1227 | |||
M32220.1 | - | 1 | 148 |