Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012276.3 | NP_001012276.2 | 3 | 471 | ||
XM_017001269.1 | XP_016856758.1 | 2 | 467 | |||
GenBank mRNA | BC140755.1 | - | 3 | 471 | 9021623 | |
BC144565.1 | - | 2 | 473 | 9053093 |
Gene Symbol: PRAMEF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012277.4 | NP_001012277.2 | 3 | 471 |