Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 15 | 2005 | ||
XM_011511216.2 | XP_011509518.1 | 11 | 1729 | |||
XM_017004159.1 | XP_016859648.1 | 11 | 1760 | |||
XM_017004160.1 | XP_016859649.1 | 7 | 728 | |||
XM_017004161.1 | XP_016859650.1 | 17 | 2442 | |||
GenBank mRNA | AY462868.1 | - | 12 | 1844 | ||
AY462871.1 | - | 15 | 2005 | |||
AY462872.1 | - | 14 | 1963 | |||
AY462873.1 | - | 13 | 1889 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 17 | 2353 | ||
XM_017004833.1 | XP_016860322.1 | 15 | 2128 | |||
XM_017004834.1 | XP_016860323.1 | 10 | 850 | |||
XM_017004835.1 | XP_016860324.1 | 8 | 787 | |||
GenBank mRNA | EF523384.1 | - | 17 | 2355 |
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277406.1 | NP_001264335.1 | 15 | 1953 | ||
XM_011511670.2 | XP_011509972.1 | 15 | 2125 | |||
XM_017004732.1 | XP_016860221.1 | 16 | 2104 | |||
XM_017004733.1 | XP_016860222.1 | 10 | 850 | |||
XM_017004734.1 | XP_016860223.1 | 8 | 787 |