Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017991.2 | NP_001017991.1 | 1 | 256 | ||
GenBank mRNA | HY000925.1 | - | 1 | 257 |
Gene Symbol: H2AFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017990.1 | NP_001017990.1 | 1 | 183 | ||
GenBank mRNA | BC128034.1 | - | 1 | 175 | 40021540 | |
CR542230.1 | - | 1 | 175 |
Gene Symbol: H2AFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080720.1 | NP_542451.1 | 1 | 183 | ||
GenBank mRNA | AF254576.1 | - | 1 | 183 | ||
BC101409.1 | - | 1 | 175 | 40021547 | ||
BC101415.1 | - | 1 | 175 | 40021541 | ||
BC101417.1 | - | 1 | 175 | 40021543 | ||
BC101418.1 | - | 1 | 175 | 40021544 | ||
BC130510.1 | - | 1 | 251 | 40146541 | ||
BC130512.1 | - | 1 | 251 | 40146543 | ||
BC134365.1 | - | 1 | 178 | 40021542 |