Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008274.3 | NP_001008275.2 | 4 | 989 | ||
XM_017023254.1 | XP_016878743.1 | 3 | 525 | |||
GenBank mRNA | AK090695.1 | - | 3 | 434 | ||
AK131485.1 | - | 4 | 934 | |||
BC033211.1 | - | 4 | 919 | 4906353 | ||
DA058421.1 | - | 3 | 434 |