Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LURAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013615.2 | NP_001013633.1 | 2 | 918 | ||
GenBank mRNA | AK057892.1 | - | 2 | 918 | ||
BX370151.2 | - | 1 | 238 |