Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012392.3 | NP_036524.1 | 3 | 756 | ||
NR_033686.1 | 2 | 455 | ||||
NR_033688.1 | 3 | 597 | ||||
XM_011541745.1 | XP_011540047.1 | 3 | 383 | |||
XM_011541746.2 | XP_011540048.1 | 3 | 850 | |||
XM_011541747.1 | XP_011540049.1 | 3 | 725 | |||
XM_017001680.1 | XP_016857169.1 | 3 | 714 | |||
XM_017001681.1 | XP_016857170.1 | 3 | 2021 | |||
GenBank mRNA | AB018357.1 | - | 3 | 395 | ||
AB026628.1 | - | 3 | 395 | |||
AK001420.1 | - | 3 | 408 | |||
AY359011.1 | - | 3 | 399 | |||
BC002773.2 | - | 3 | 385 | 3615612 | ||
BC012561.1 | - | 3 | 401 | 4273213 | ||
BG546473.1 | - | 2 | 115 | |||
BI826532.1 | - | 3 | 569 | |||
CR542139.1 | - | 3 | 383 |