Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048252.2 | NP_001041717.1 | 3 | 662 | ||
NM_001127385.1 | NP_001120857.1 | 3 | 471 | |||
GenBank mRNA | AB219764.1 | - | 3 | 639 | ||
AB219832.1 | - | 3 | 437 | |||
BC071790.1 | - | 3 | 435 | |||
BC147012.1 | - | 1 | 114 | 9056940 |